Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Fluoro-4-nitrobenzoic acid, 95+%, Thermo Scientific™

Catalog No. TL00808DA
Click to view available options
Quantity:
1 g
Packaging:
Amber Glass Bottle

Chemical Identifiers

CAS 403-24-7
Molecular Formula C7H4FNO4
Molecular Weight (g/mol) 185.11
InChI Key MMWFMFZFCKADEL-UHFFFAOYSA-N
Synonym 2-fluoro-4-nitrobenzoic acid, 2-fluoro-4-nitrobenzenecarboxylic acid, 2-fluoro-4-nitrobenzoicacid, 2-fluoro-4-nitro-benzoic acid, 4-carboxy-3-fluoronitrobenzene, benzoic acid, 2-fluoro-4-nitro, pubchem1310, acmc-1cts9, 2-fluoro4-nitrobenzoic acid, 2-fluor-4-nitro-benzoesaure
PubChem CID 302679
SMILES C1=CC(=C(C=C1[N+](=O)[O-])F)C(=O)O

Specifications

Assay Percent Range 95%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 185.11
Percent Purity 95%
Chemical Name or Material 2-Fluoro-4-nitrobenzoic acid
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.