Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
2-Hexyl-1-n-octanol 98.0+%, TCI America™
Shop All Tokyo Chemical Industry (TCI) Products

Click to view available options
Quantity:
1 mL
5 mL
Specifications
Specifications
Color | Yellow |
Boiling Point | 162°C |
Quantity | 1 mL |
Formula Weight | 214.39 |
Percent Purity | ≥98.0% (GC) |
Physical Form | Liquid |
Chemical Name or Material | 2-Hexyl-1-n-octanol |
Safety and Handling
EINECSNumber | (2)-0217&(2)-3704 |
TSCA | No |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction