Learn More
TARGETMOL CHEMICALS INC 2-HYDROXY ATORVASTATIN C 5MG
Also available in 1 mg 50 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. 2-Hydroxy atorvastatin calcium salt is a hydroxy metabolite of Atorvastatin calcium salt which is a potent HMG-CoA reductase inhibitor (IC50 = 8 nM). purity: 97%