Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Indancarboxylic Acid 98.0+%, TCI America™
SDP

Catalog No. p-7131464 Shop All Tokyo Chemical Industry (TCI) Products
Click to view available options
Quantity:
1 g
5 g

Chemical Identifiers

CAS 25177-85-9
Molecular Formula C10H10O2
Molecular Weight (g/mol) 162.19
MDL Number MFCD00085095
InChI Key XUDCMQBOWOLYCF-UHFFFAOYSA-N
Synonym 2-indancarboxylic acid, indane-2-carboxylic acid, indan-2-carboxylic acid, 2-indanecarboxylic acid, 1h-indene-2-carboxylic acid, 2,3-dihydro, chembl81558, 2-carboxyindane, 2-carboxy-indan, 2-carboxy-indane, 2-indancarboxylicacid
PubChem CID 575777
IUPAC Name 2,3-dihydro-1H-indene-2-carboxylic acid
SMILES OC(=O)C1CC2=CC=CC=C2C1

Specifications

Melting Point 128°C
Boiling Point 192°C
Quantity 1 g
Formula Weight 162.19
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 2-Indancarboxylic Acid
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.