Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Iodo-5-methylaniline 98.0+%, TCI America™
SDP

Catalog No. p-7131723 Shop All Tokyo Chemical Industry (TCI) Products
Click to view available options
Quantity:
1 g
5 g

Chemical Identifiers

CAS 13194-69-9
Molecular Formula C7H8IN
Molecular Weight (g/mol) 233.052
MDL Number MFCD00833395
InChI Key KXPBTNCFONSVIA-UHFFFAOYSA-N
Synonym 3-Amino-4-iodotoluene
PubChem CID 2736659
IUPAC Name 2-iodo-5-methylaniline
SMILES CC1=CC(=C(C=C1)I)N

Specifications

Melting Point 36°C
Quantity 1 g
Formula Weight 233.05
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2-Iodo-5-methylaniline
TSCA No
Recommended Storage Refrigerator
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.