Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Iodohippuric Acid 99.0+%, TCI America™
SDP

Catalog No. p-7131411 Shop All Tokyo Chemical Industry (TCI) Products
Click to view available options
Quantity:
5 g
25 g

Chemical Identifiers

CAS 147-58-0
Molecular Formula C9H8INO3
Molecular Weight (g/mol) 305.071
MDL Number MFCD00002694
InChI Key CORFWQGVBFFZHF-UHFFFAOYSA-N
Synonym N-(2-Iodobenzoyl)glycine
PubChem CID 8614
IUPAC Name 2-[(2-iodobenzoyl)amino]acetic acid
SMILES C1=CC=C(C(=C1)C(=O)NCC(=O)O)I

Specifications

Melting Point 173°C
Color White
Quantity 5 g
Formula Weight 305.07
Percent Purity ≥99.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 2-Iodohippuric Acid
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.