Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
2-Iodomelatonin, Tocris Bioscience™
Shop All R&D Systems Products
Click to view available options
Quantity:
10 mg
50 mg
Description
Potent melatonin agonist (pKi values are 10.55 and 9.87 at human recombinant MT1 and MT2 receptors respectively). 2-[125I]-iodomelatonin has been used to identify, characterize and localize melatonin binding sites in the brain and peripheral tissues.
Specifications
Specifications
Quantity | 10 mg |
Formula Weight | 358.18 |
Percent Purity | >98% |
Chemical Name or Material | 2-Iodomelatonin |
Safety and Handling
Recommended Storage | Desiccate at +4°C |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction