Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
2-Isobutyl-3-methoxypyrazine 97.0+%, TCI America™
Shop All Tokyo Chemical Industry (TCI) Products

Click to view available options
Quantity:
1 mL
5 mL
Specifications
Specifications
Color | Yellow |
Quantity | 1 mL |
Formula Weight | 166.22 |
Percent Purity | ≥97.0% (GC,T) |
Physical Form | Liquid |
Chemical Name or Material | 2-Isobutyl-3-methoxypyrazine |
Safety and Handling
TSCA | Yes |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction