Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Isopropylphenyl isothiocyanate, ≥95%, Thermo Scientific™

Catalog No. SB01626EB
Click to view available options
Quantity:
25 g

Chemical Identifiers

CAS 36176-31-5
Molecular Formula C10H11NS
Molecular Weight (g/mol) 177.265
MDL Number MFCD00022053
InChI Key QQOOMDSEMQHMIR-UHFFFAOYSA-N
Synonym 2-isopropylphenyl isothiocyanate, 1-isopropyl-2-isothiocyanatobenzene, 1-isothiocyanato-2-propan-2-yl benzene, benzene, 1-isothiocyanato-2-1-methylethyl, 2-methylethyl benzenisothiocyanate, 2-isopropylphenylisothiocyanate, acmc-20ansf, 2-isopropylphenyl-isothiocyanate, 2-iso-propylphenyl isothiocyanate
PubChem CID 142052
IUPAC Name 1-isothiocyanato-2-propan-2-ylbenzene
SMILES CC(C)C1=CC=CC=C1N=C=S

Specifications

CAS Min % 95.0
Assay Percent Range 95%+
Linear Formula CH3CH(CH3)C6H4NCS
Quantity 25 g
Formula Weight 177.27
Percent Purity 95%
Chemical Name or Material 2-Isopropylphenyl isothiocyanate
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Harmful if swallowed.
May cause respiratory irritation.
Causes serious eye irritation.
Precautionary Statement GHS P Statement:
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.