Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Keto-D-gluconic acid, hemicalcium salt monohydrate, >98%, MP Biomedicals™

Catalog No. ICN10036510 Shop All MP Biomedicals, Inc Products
Click to view available options
Quantity:

2-Keto-D-gluconic Acid hemicalcium salt is Hemicalcium Salt of 2-Keto-D-gluconic acid. It is produced from D-glucose

Chemical Identifiers

CAS

3470-37-9

Molecular Formula

C12H20CaO14

Molecular Weight (g/mol)

428.356

MDL Number

MFCD00149104

InChI Key

QACANIDDQFGAJU-JQTJLMCZSA-N

Synonym

calcium 2-keto-d-gluconate

PubChem CID

131852723

IUPAC Name

calcium;(3S,4R,5R)-3,4,5,6-tetrahydroxy-2-oxohexanoic acid

SMILES

C(C(C(C(C(=O)C(=O)O)O)O)O)O.C(C(C(C(C(=O)C(=O)O)O)O)O)O.[Ca]

Specifications

Color

White

Assay Percent Range

>98%

Quantity

10 g

Formula Weight

213.2

Physical Form

Powder

Chemical Name or Material

2-Keto-D-gluconic acid, hemicalcium salt

Recommended Storage

Store at +4°C.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.