Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Mercapto-4(3H)-quinazolinone 98.0+%, TCI America™
SDP

Catalog No. p-7132532 Shop All Tokyo Chemical Industry (TCI) Products
Click to view available options
Quantity:
5 g
25 g

Chemical Identifiers

CAS 13906-09-7
Molecular Formula C8H6N2OS
Molecular Weight (g/mol) 178.209
MDL Number MFCD00006885
InChI Key PUPFOFVEHDNUJU-UHFFFAOYSA-N
Synonym 2-Thio-4(3H)-quinazolinone
PubChem CID 683940
IUPAC Name 2-sulfanylidene-1H-quinazolin-4-one
SMILES C1=CC=C2C(=C1)C(=O)NC(=S)N2

Specifications

Color White
Quantity 5 g
Formula Weight 178.21
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2-Mercapto-4(3H)-quinazolinone
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.