Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Mercaptoacetamide (in Methanolic Ammonia Solution 10g/100ml approx.), 97%, Thermo Scientific™

Catalog No. GK00437EA
Click to view available options
Quantity:
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS

758-08-7, 7664-41-7, 67-56-1

Molecular Formula

C2H5NOS

Molecular Weight (g/mol)

91.128

MDL Number

MFCD00068159

InChI Key

GYXHHICIFZSKKZ-UHFFFAOYSA-N

Synonym

2-mercaptoacetamide, mercaptoacetamide, thioglycolamide, acetamide, 2-mercapto, thioglycolamide crude, usaf ha-3, acetamide, 2-mercapto-6ci,7ci,8ci,9ci, 2-mercaptoacetamide in solution nh3/meoh, thiolacetamide, a-mercaptoacetamide

PubChem CID

12961

IUPAC Name

2-sulfanylacetamide

SMILES

C(C(=O)N)S

Specifications

CAS Min %

10.0, 7.0, 75.0

CAS Max %

12.0, 13.0, 83.0

Assay Percent Range

97%

Linear Formula

SHCH2C(O)NH2

Quantity

10 g

Formula Weight

91.13

Percent Purity

97%

Chemical Name or Material

2-Mercaptoacetamide (in Methanolic Ammonia Solution 10g/100ml approx.)

Hazard Category

Danger

Hazard Statement

GHS P Statement Causes damage to organs. Causes severe skin burns and eye damage. Toxic if swallowed. Toxic in contact with skin. Fatal if inhaled. Highly flammable liquid and vapor.

Precautionary Statement

GHS P Statement Wear protective gloves/protective clothing/eye protection/face protection. IF SWALLOWED: rinse mouth. Do NOT induce vomiting. Immediately call a POISON CENTER or doctor/physician. IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/shower. IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing. Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.