Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Mercaptoacetamide (in Methanolic Ammonia Solution 10g/100ml approx.), 97%, Thermo Scientific™

Catalog No. p-2698544
Click to view available options
Quantity:
1 g
10 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 758-08-7, 7664-41-7, 67-56-1
Molecular Formula C2H5NOS
Molecular Weight (g/mol) 91.128
MDL Number MFCD00068159
InChI Key GYXHHICIFZSKKZ-UHFFFAOYSA-N
Synonym 2-mercaptoacetamide, mercaptoacetamide, thioglycolamide, acetamide, 2-mercapto, thioglycolamide crude, usaf ha-3, acetamide, 2-mercapto-6ci,7ci,8ci,9ci, 2-mercaptoacetamide in solution nh3/meoh, thiolacetamide, a-mercaptoacetamide
PubChem CID 12961
IUPAC Name 2-sulfanylacetamide
SMILES C(C(=O)N)S

Specifications

CAS Min % 10.0, 7.0, 75.0
CAS Max % 12.0, 13.0, 83.0
Assay Percent Range 97%
Linear Formula SHCH2C(O)NH2
Quantity 1 g
Formula Weight 91.13
Percent Purity 97%
Chemical Name or Material 2-Mercaptoacetamide (in Methanolic Ammonia Solution 10g/100ml approx.)
Hazard Category Danger
Hazard Statement GHS P Statement Causes damage to organs. Causes severe skin burns and eye damage. Toxic if swallowed. Toxic in contact with skin. Fatal if inhaled. Highly flammable liquid and vapor.
Precautionary Statement GHS P Statement Wear protective gloves/protective clothing/eye protection/face protection. IF SWALLOWED: rinse mouth. Do NOT induce vomiting. Immediately call a POISON CENTER or doctor/physician. IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/shower. IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing. Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.