Learn More
ILEX LIFE SCIENCES LLC 2-MERCAPTOETHANOL 50MM 100ML
Supplier: ILEX LIFE SCIENCES LLC P0705100
PAN-Biotech 2-Mercaptoethanol 50 mM in DPBS (100 ml). DESCRIPTION: 2-Mercaptoethanol, also known as beta-mercaptoethanol (BME), is a potent reducing agent used in cell culture medium to prevent toxic levels of oxygen radicals. 2-Mercaptoethanol is not stable in solution, thus protocols generally require daily supplementation. PAN-Biotech 2-Mercaptoethanol is supplied at a concentration of 50 mM in Dulbecco's Phosphate Buffered Saline (DPBS). SOLUTION: 50 mM in DPBS. STORAGE: +2-8°C. STERILE: Yes. MANUFACTURER: PAN-Biotech GmbH. COUNTRY OF ORIGIN: Germany.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.