Learn More
ILEX LIFE SCIENCES LLC 2-MERCAPTOETHANOL 50MM 20ML
SDP
Supplier:  ILEX LIFE SCIENCES LLC P0705020
Encompass_Preferred
PAN-Biotech 2-Mercaptoethanol 50 mM in DPBS (20 ml). DESCRIPTION: 2-Mercaptoethanol, also known as beta-mercaptoethanol (BME), is a potent reducing agent used in cell culture medium to prevent toxic levels of oxygen radicals. 2-Mercaptoethanol is not stable in solution, thus protocols generally require daily supplementation. PAN-Biotech 2-Mercaptoethanol is supplied at a concentration of 50 mM in Dulbecco's Phosphate Buffered Saline (DPBS). SOLUTION: 50 mM in DPBS. STORAGE: +2-8°C. STERILE: Yes. MANUFACTURER: PAN-Biotech GmbH. COUNTRY OF ORIGIN: Germany.
Catalog No. 50-273-3717
missing translation for 'QuantityLeft'
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.