Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Methoxyethyl Acetoacetate 95.0+%, TCI America™
SDP

Catalog No. p-7117532 Shop All Tokyo Chemical Industry (TCI) Products
Click to view available options
Quantity:
25 g
500 g

Chemical Identifiers

CAS 22502-03-0
Molecular Formula C7H12O4
Molecular Weight (g/mol) 160.17
MDL Number MFCD00009648
InChI Key PLHCSZRZWOWUBW-UHFFFAOYSA-N
Synonym 2-methoxyethyl acetoacetate, 2-methoxyethylacetoacetate, acetoacetic acid 2-methoxyethyl ester, butanoic acid, 3-oxo-, 2-methoxyethyl ester, methoxyethyl acetoacetate, acmc-1cs35, 2-methoxyethyl-3-oxobutanoate, #, 2-methoxy ethyl acetoacetate, 2-methoxyethyl acetoacetate, lonza quality gc
PubChem CID 89734
IUPAC Name 2-methoxyethyl 3-oxobutanoate
SMILES COCCOC(=O)CC(C)=O

Specifications

Color Yellow
Boiling Point 120°C
Quantity 25 g
Formula Weight 160.17
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Methoxyethyl Acetoacetate
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.