Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Methylpyrrolidine hydrochloride, 97%, Thermo Scientific™

Catalog No. AC458070050
Click to view available options
Quantity:

Chemical Identifiers

CAS

54677-53-1

Molecular Formula

C5H11N·HCl

Molecular Weight (g/mol)

121.61

InChI Key

JNEIFWYJFOEKIM-UHFFFAOYSA-N

Synonym

2-methylpyrrolidine hydrochloride, 2-methylpyrrolidine hcl, 2-methyl-pyrrolidine hydrochloride, pubchem11112, pubchem11115, acmc-1c01e, 2-methylpyrrolidinehydrochloride, 2-methylpyrrolidine-hydrogen chloride 1/1, pyrrolidine, 2-methyl-, hydrochloride 1:1, pyrrolidine, 2-methyl-, hydrochloride, 2r-9ci

PubChem CID

21907341

IUPAC Name

2-methylpyrrolidine;hydrochloride

SMILES

CC1CCCN1.Cl

Specifications

Infrared Spectrum

Authentic

Quantity

5 g

Solubility Information

Solubility in water: slightly soluble.

Formula Weight

121.61

Percent Purity

97%

Chemical Name or Material

2-Methylpyrrolidine hydrochloride

Hazard Category

GHS Signal Word: Warning

Hazard Statement

GHS H Statement
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

Precautionary Statement

GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and kee

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.