Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-(Methylsulfonyl)phenol, 98%, Thermo Scientific Chemicals

Catalog No. p-7077577
Click to view available options
Quantity:
1 g
5 g

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 27489-33-4
Molecular Formula C7H8O3S
Molecular Weight (g/mol) 172.2
InChI Key ZWTZHHHSRDUVRT-UHFFFAOYSA-N
PubChem CID 598998
IUPAC Name 2-methylsulfonylphenol
SMILES CS(=O)(=O)C1=CC=CC=C1O

Specifications

Melting Point 88.0°C to 90.0°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Packaging Glass Bottle
Linear Formula HOC6H4SO2CH3
Quantity 1 g
Synonym 2-methylsulfonyl phenol, 2-methanesulfonylphenol, 2-methylsulfonyl benzenol, phenol, 2-methylsulfonyl, phenol,2-methylsulfonyl, phenol, o-methylsulfonyl, methylsulfonylphenol, methylsulfonylbenzenol, acmc-1cbus, 2-methanesulfonyl-phenol
Formula Weight 172.2
Percent Purity 98%
Chemical Name or Material 2-(Methylsulfonyl)phenol
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.