Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Nitro-p-tolunitrile 98.0+%, TCI America™
SDP

Catalog No. p-7133837 Shop All Tokyo Chemical Industry (TCI) Products
Click to view available options
Quantity:
1 g
5 g

Chemical Identifiers

CAS 26830-95-5
Molecular Formula C8H6N2O2
Molecular Weight (g/mol) 162.15
MDL Number MFCD00056107
InChI Key QGBSLPHQCUIZKK-UHFFFAOYSA-N
Synonym 2-nitro-p-tolunitrile, 2-nitro-4-toluonitrile, benzonitrile, 4-methyl-2-nitro, 2-nitro-4-methylbenzonitrile, 4-methyl-2-nitro-benzonitrile, 4-methyl-2-nitrobenzenecarbonitrile, pubchem13730, acmc-1cjm6, ksc494k5h, 4-methyl-2-nitro benzonitrile
PubChem CID 96235
IUPAC Name 4-methyl-2-nitrobenzonitrile
SMILES CC1=CC=C(C#N)C(=C1)[N+]([O-])=O

Specifications

Melting Point 99°C
Color Yellow
Quantity 1 g
Formula Weight 162.15
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-Nitro-p-tolunitrile
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.