Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Phenylbenzoxazole 98.0+%, TCI America™
SDP

Catalog No. p-7135485 Shop All Tokyo Chemical Industry (TCI) Products
Click to view available options
Quantity:
1 g
5 g

Chemical Identifiers

CAS 833-50-1
Molecular Formula C13H9NO
Molecular Weight (g/mol) 195.221
MDL Number MFCD00012183
InChI Key FIISKTXZUZBTRC-UHFFFAOYSA-N
Synonym 2-phenylbenzoxazole, 2-phenylbenzo d oxazole, benzoxazole, 2-phenyl, ccris 7874, phenyl-2 benzoxazole french, 2-phenyl-benzoxazole, phenyl-2 benzoxazole, 2-phenybenzoxazole, 2-phenyl benzoxazole, 2-phenyl-benzooxazole
PubChem CID 70030
IUPAC Name 2-phenyl-1,3-benzoxazole
SMILES C1=CC=C(C=C1)C2=NC3=CC=CC=C3O2

Specifications

Melting Point 103°C
Quantity 1 g
Formula Weight 195.22
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-Phenylbenzoxazole
RTECSNumber DM4865700
TSCA Yes
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.