Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Propyl-2-imidazoline, TCI America™
SDP

Catalog No. P116425G Shop All Tokyo Chemical Industry (TCI) Products
Click to view available options
Quantity:
25 g

Chemical Identifiers

CAS 15450-05-2
Molecular Formula C6H12N2
Molecular Weight (g/mol) 112.176
MDL Number MFCD00191487
InChI Key LFCYHAXVBWPGQR-UHFFFAOYSA-N
PubChem CID 27291
IUPAC Name 2-propyl-4,5-dihydro-1H-imidazole
SMILES CCCC1=NCCN1

Specifications

Melting Point 42°C
Color Yellow
Boiling Point 140°C
Quantity 25 g
Formula Weight 112.18
Physical Form Crystalline Powder
Chemical Name or Material 2-Propyl-2-imidazoline
RTECSNumber NJ4425000
TSCA No
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.