Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
2-pyrrolidin-1-ylaniline, Thermo Scientific™
Click to view available options
Quantity:
250 mg
1 g
5 g
Specifications
Specifications
Assay Percent Range | 97% |
Quantity | 250 mg |
Formula Weight | 162.23 |
Percent Purity | 97% |
Chemical Name or Material | 2-Pyrrolidin-1-ylaniline |
Safety and Handling
Hazard Category | GHS Signal Word: Warning |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction