Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(2-Pyrrolidin-1-ylphenyl)methanol, 95%, Thermo Scientific™

Catalog No. p-4511327
Click to view available options
Quantity:
250 mg
1 g
5 g
Packaging:
Amber Glass Bottle

Chemical Identifiers

CAS 73051-88-4
Molecular Formula C11H15NO
Molecular Weight (g/mol) 177.247
InChI Key QXIYEQQTAPJDBX-UHFFFAOYSA-N
Synonym 2-pyrrolidin-1-ylphenyl methanol, 2-pyrrolidin-1-yl phenyl methanol, 2-pyrrolizinobenzenemethanol, 2-1-pyrrolidinyl benzylalkohol, 2-pyrrolidinylphenyl methan-1-ol, benzenemethanol,2-1-pyrrolidinyl
PubChem CID 13021765
IUPAC Name (2-pyrrolidin-1-ylphenyl)methanol
SMILES C1CCN(C1)C2=CC=CC=C2CO

Specifications

Assay Percent Range 95%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 177.25
Percent Purity 95%
Chemical Name or Material (2-Pyrrolidin-1-ylphenyl)methanol
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.