Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-(tert-Butyl)-4,6-dimethylphenol, Tech., Thermo Scientific™

Catalog No. SB01223EA
Click to view available options
:
Amber Glass Bottle; 10g

Specifications

CAS 1879-09-0
Molecular Formula C12H18O
Linear Formula (CH3)2(OH)C6H2C(CH3)3
MDL Number MFCD00002234
Synonym 2,4-dimethyl-6-tert-butylphenol, 2-tert-butyl-4,6-dimethylphenol, 6-tert-butyl-2,4-dimethylphenol, topanol a, 6-tert-butyl-2,4-xylenol, 6-t-butyl-2,4-xylenol, prodox 340, phenol, 2-1,1-dimethylethyl-4,6-dimethyl, 6-t-butyl-2,4-dimethylphenol, 2,4-xylenol, 6-tert-butyl
InChI Key OPLCSTZDXXUYDU-UHFFFAOYSA-N
SMILES CC1=CC(C)=C(O)C(=C1)C(C)(C)C
IUPAC Name 2-tert-butyl-4,6-dimethylphenol
Molecular Weight (g/mol) 178.28
PubChem CID 15884
Formula Weight 178.27
Percent Purity Tech.
Grade Technical
Chemical Name or Material 2-(tert-Butyl)-4, 6-dimethylphenol
Show More Show Less
Hazard Statement GHS H Statement:
May cause respiratory irritation.
Harmful if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.