Learn More
Accuflow Systems Accuflow Systems Sterile Empty Vial (SEV), 20 mL, 20 mm glass with silver cap - 16/Pack
Supplier:  Accuflow Systems SEV2016
Encompass Procurement Services
Non-distribution item offered as a customer accommodation; additional freight charges may apply.
Learn More
Encompass_Preferred
ALK Sterile Empty Vials (SEV) feature Type 1 glass USP borosilicate, and a latex free stopper. ALK SEVs are produced at a cGMP FDA Manufacturer and USP tested for sterility and endotoxins. Certificate of Analysis available for all production lots and vials are aseptically assembled in an ISO 5 (class 100) cleanroom. 16/Pack
Catalog No. NC1741086
missing translation for 'QuantityLeft'