Learn More
Ace Glass, Inc. 2000mL graduated pressure equalizing funnel, 29/42 top, PTFE metering plug
Supplier:  Ace Glass, Inc. 732017
Encompass_Preferred
2000ml graduated cylindrical pressure equalizing funnel with 29/42 top and bottom joints, drip stem, pressure equalizing arm, stopcock w/PTFE metering plug, and double scale graduations calibrated from stopcock.
Catalog No. 50-974-788
missing translation for 'QuantityLeft'
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.