Learn More
Ace Glass, Inc. 2000mL graduated pressure equalizing funnel, 29/42 top, glass plug
Supplier:  Ace Glass, Inc. 729658
Encompass_Preferred
2000mL graduated pressure equalizing funnel featuring a 29/42 standard taper joint at top. Cylindrical funnel features standard taper joints at the top and bottom, pressuring equalizing arm and double scale graduations calibrated from stopcock. Also featured is a drip system that does not extend beyond the joint to eliminate possible sources of breakage. Glass plug included.
Catalog No. 50-974-774
missing translation for 'QuantityLeft'
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.