Learn More
Nanocomposix BioReady Magnetic Gold Nanoshells - Carboxyl - 200 nm, 10 OD in aqueous 0.02 mM potassium carbonate, 1 mL
As a global leader in gold nanoparticle production, nanoComposix specializes in providing high-purity and monodisperse nanomaterials – delivering the quality you need for consistent results.200 nm magnetic gold nanoshells combine the plasmonic properties of gold nanoparticles with the separation capability of a magnetic bead. The magnetic core allows for easy manipulation such as separation or concentration and enhances particle utility for a variety of therapeutic and diagnostic applications such as biosensors, lateral flow assays, SERS, drug delivery, and photothermal therapy.BioReady Magnetic Gold Nanoshells offer a carboxyl ligand (lipoic acid) that is covalently bound to the metal surface enabling subsequent covalent conjugation. Antibodies and other amine-containing biomolecules efficiently bind to carboxyl gold nanoshells using EDC-NHS chemistry to make highly stable conjugates. Review our conjugation protocol or contact our experts for support.