Learn More
Ace Glass, Inc. 220mm A quartz immersion well, Tube connection
Supplier:  Ace Glass, Inc. 785425
Encompass_Preferred
220mm A double-walled quartz immersion well with inlet and outlet tubes for cooling. Inlet tube extends down the annular space to insure flow of coolant from bottom of well upward to outlet. Well has 60/40 inner joint. O.D. of outer jacket is 53mm max. Length above joint is 76mm.
Catalog No. 50-979-494
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.