Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SHOWA™ 234 HPPE Cut-Resistant Gloves, Foam Nitrile Coating

Catalog No. p-7156822 Shop All Showa Best Glove Products
Click to view available options
Size:
Small
Medium
Large
X-Large
This item is not returnable. View return policy
This item is not returnable. View return policy

Designed for use in 'knife hand' applications in food processing, these flexible cut protection gloves combine foam nitrile with a HPPE liner for protection in both dry and greasy environments.

  • 15-gauge seamless knit spandex liner with HPPE
  • Foamed flat dipped coating with sponge finish for secure grip
  • Contact Heat level 2
  • FDA and EU Food Contact approved
  • Lightweight, supple liner with breathable open back design
  • Foam nitrile coating protects against oils, hydrocarbons, grease and abrasions
  • Cooling HPPE properties and breathable back of hand reduces perspiration and keeps hands dry
  • Thin and lightweight for enhanced dexterity and longer use
  • Ergonomically designed to replicate the hand’s natural curves
  • Launderable for re-use, leads to less waste and cost efficiency
  • Seamless knit designed to prevent irritation

Specifications

Certifications/Compliance ANSI/ISEA 105-16: Level A4 (minimum 1500g)
Certifications/Compliance (Europe) EN 388:2016 Cut Level D
Construction Seamless Knit
Coating Material Sponge Nitrile
Color Gray, Black
Cut Resistance Rating ANSI A4
Latex Free No
Material HPPE, Spandex
Product Type Cut-resistant Gloves
Size Medium
EN388 Standard EN 388:4X42D
For Use With (Application) HVAC, maintenance, mechanical and engineering, steel fabrication, washing and cleaning, component assembly
Liner Material HPPE
Model 234M
Series 234 Series
Yarn Gauge 15
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.