Learn More
Enzo Life Sciences 24(R),25-Dihydroxyvitamin D3 (50µg). CAS: 55721-11-4
SDP
Supplier:  Enzo Life Sciences BMLDM3000050
Encompass_Preferred
Major metabolite of vitamin D3. Mediates calcium and phosphorous homeostasis. Inhibits 1α,25-dihydroxyvitamin D3 and testosterone potentiation of calcium channels in osteosarcoma cells. Formulation: White solid dissolved in ethanol (50µg/50µl). Purity: ≥98% (TLC). Long Term Storage: -80°C.
Catalog No. 50-201-0517
missing translation for 'QuantityLeft'
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.