Learn More
Cytiva (Formerly Pall Lab) AcroPrep 24-well Sterile Filtration Plate-0.65/0.2 m Supor EKV (8/pkg)
Supplier:  Cytiva 97027
Encompass Procurement Services
Non-distribution item offered as a customer accommodation; additional freight charges may apply.
Learn More
Encompass_Preferred
AcroPrep 24-well Sterile Filtration Plate-0.65/0.2 m Supor EKV (2/pkg). Palls sterile filtration AcroPrep 24-well filter plate iswell suited for high volume (up to 7 mL) plate-basedsterile filtration needs such as media, reagent, serumor proteins. Compatible with either a vacuum manifoldor centrifuge, the 24-well Supor EKV plate features adual-layer, 0.65 m membrane integrated with a highlyasymmetric 0.2 m membrane for fast, efficient, andsterilizing grade filtration.
Catalog No. NC1857355
missing translation for 'QuantityLeft'