Learn More
Electron Microscopy Sciences MAC 25 Thin Foil Standards for STEM
Supplier:  Electron Microscopy Sciences 8008225
Encompass Procurement Services
Non-distribution item offered as a customer accommodation; additional freight charges may apply.
Learn More
Encompass_Preferred
We have a selection of 25 high purity thin foils, 0.1 mm thick, 3 mm diameter to fit into TEM grid holders for use in the STEM mode. They are polished to a 3 micron diamond finish, and come in a variety of pure metals: Aluminum, Cadmium, Cobalt, Copper, Gold, Hafnium, Indium, Iron, Magnesium, Molybdenum, Nickel, Niobium, Palladium, Platinum, Rhodium, Silver, Tantalum, Tin, Titanium, Tungsten, Vanadium, Yttrium, Zinc, Zirconium, AISI 316 S/S
Catalog No. 50-293-04