Learn More
Chemglass Life Sciences 25mL Peptide Synthesis Vessel, Solid Phase, Medium Frit, GL 25 Thread
SDP
Supplier:  Chemglass Life Sciences CG186002
Encompass_Preferred
25mL Peptide Synthesis Vessel, Solid Phase, Medium Frit, GL 25 Thread. Solid phase peptide synthesis vessel having a medium or coarse porosity fritted glass resin support. Top of the vessel has a GL thread and is supplied complete with a PTFE lined PBT screw cap. Lower PTFE stopcock has a 2mm bore with the arm being 8mm O.D. Vessels are available in sizes other than those listed. Please call for details.
Catalog No. CG186002
missing translation for 'QuantityLeft'
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.