Learn More
Chemglass Life Sciences 25mL Addition Funnel, Graduated, 14/20 Joint Size, 2mm PTFE Stpk, 195mm OAH
Supplier: Chemglass Life Sciences CG171813
25mL Addition Funnel, Graduated, 14/20 Joint Size, 2mm PTFE Stpk, 195mm OAH. Cylindrical style addition funnel similar to CG-1716, but with PTFE stopcocks. Funnels with 14/20 and 19/22 joints are supplied complete with a stopper. For replacement stoppers see CG-3000.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.