Learn More
Ace Glass, Inc. 25ML VOLUMETRIC BULB 25ML VOLUMETRIC BULB
Supplier:  Ace Glass, Inc. 800436
Encompass_Preferred
25mL volumetric bulb for use with ACE Automatic Dispensing Pipet. Bulbs have a short tipping angle and give reproduceable volumes.
Catalog No. 50-975-875
missing translation for 'QuantityLeft'
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.