Learn More
Cell Biolabs Inc 293AAV Cell Line for AAV Packaging
Supplier:  Cell Biolabs Inc AAV100
Encompass Procurement Services
Non-distribution item offered as a customer accommodation; additional freight charges may apply.
Learn More
Encompass_Preferred
Cell Biolabs' 293AAV Cell Line is a permanent line established from primary embryonic human kidney transformed with human adenovirus type 5 DNA. The genes encoded by the E1 region of adenovirus (E1a and E1b) are expressed in these cells and participate in transactivation of viral promoters, allowing these cells to produce high levels of protein. The 293AAV Cell Line is derived from the parental 293 cells but selected for attributes that increase AAV production. It offers several advantages over the regular 293 cells: (1) Larger cell surface area resulting higher transfection and better yield of AAV; (2) Flattened morphology; and (3) Firm attachment to culture plates, ideal for large scale culture and AAV production.
Catalog No. 50-672-451
missing translation for 'QuantityLeft'