Learn More
Ace Glass, Inc. 2L 100MMID JKTD BODY
Supplier:  Ace Glass, Inc. 6386320
Encompass Procurement Services
Non-distribution item offered as a customer accommodation; additional freight charges may apply.
Learn More
Encompass_Preferred
Heavy wall 2L jacketed filter reactor body with 100mm I.D. grooved top flange, #80 bottom thread, a CAPFE o-ring, and #11 Ace-Thred inlet/outlet connections. Dimensions: 130mm I.D. x 180mm depth. Pressure rated to 35psi at 100 Deg.C.
Catalog No. 50-978-676