Learn More
Ace Glass, Inc. 2L heavy wall filter reactor with five neck head, #80threaded body, 10mm stir shaft and bearing, and 3/8" female NPT 100 micron polyethylene filter adapter. Pressure rated at 35 psi.
Supplier:  Ace Glass, Inc. 6384239
Encompass_Preferred
Catalog No. 50-972-797
missing translation for 'QuantityLeft'
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.