Learn More
Ace Glass, Inc. 2L JKTD PRES REAC C
Supplier:  Ace Glass, Inc. 6386120
Encompass_Preferred
Heavy wall 2L jacketed filter reactor, pressure rated to 35 psi at 100 Deg.C. Has 28/15 o-ring ball jt inlet/outlets, 100mm Ace-Thred head, an 80mm btm valve adapter, filter supports, stir shaft, bearing, condenser, funnel, clamp, and o-rings.
Catalog No. 50-972-807
missing translation for 'QuantityLeft'
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.