Learn More
Thomas Scientific Aspirating Pipette, 2mL, Sterile, Individually Wrapped
SDP
Supplier:  Thomas Scientific 1196M88
Encompass
Aspirating pipettes are sterile, individually wrapped plastic serological pipettes that are supplied without filter plugs and graduations for vacuum aspirating applications. They are very useful in tissue culture applications when aspirating larger volumes from dishes, flasks, or roller bottles.
Catalog No. 50-145-8105
missing translation for 'QuantityLeft'
Explore all special offers
Add to Cart
This item is not returnable. View return policy
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.