Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(2S,3S)-(-)-Bis(diphenylphosphino)butane 93.0+%, TCI America™
SDP

Catalog No. p-7119726 Shop All Tokyo Chemical Industry (TCI) Products
Click to view available options
Quantity:
100 mg
1 g

Chemical Identifiers

CAS 64896-28-2
Molecular Formula C28H28P2
Molecular Weight (g/mol) 426.48
MDL Number MFCD00009836
InChI Key FWXAUDSWDBGCMN-ZBXWWWQKNA-N
Synonym s,s-chiraphos, 2s,3s---bis diphenylphosphino butane, chiraphos, chiraphos, s,s, unii-6qr78gzl9b, 6qr78gzl9b, 2s,3s-butane-2,3-diylbis diphenylphosphine, phosphine, 1s,2s-1,2-dimethyl-1,2-ethanediyl bis diphenyl, --chiraphos, 2s,3s-chiraphos
PubChem CID 10113249
IUPAC Name [(2S,3S)-3-(diphenylphosphanyl)butan-2-yl]diphenylphosphane
SMILES C[C@@H]([C@H](C)P(C1=CC=CC=C1)C1=CC=CC=C1)P(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point 109°C
Color White
Quantity 100 mg
Formula Weight 426.48
Percent Purity ≥93.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material (2S,3S)-(-)-Bis(diphenylphosphino)butane
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.