Learn More
Research Products International Corp 2XYT Broth, Granulated, 5 Kilograms
Supplier: Research Products International Corp X156805000.0
Nutritionally enriched medium for the growth of recombinant strains of E. Coli and in the preparation and transformation of competent cells. Material is specially granulated to reduce dust and increase solubility rate. Dissolve 31g in one liter distilled water. Heat solution to boiling with continuous stirring. Sterilize by autoclaving at 121?C for 15 minutes. Final pH@ 25?C: 6.8-7.2. Ingredients: Digest Peptone: 16 g/l Yeast Extract: 10 g/l Sodium Chloride: 5 g/l
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.