Learn More
Research Products International Corp 2XYT Broth, Powder, 1 Kilogram
SDP
Supplier:  Research Products International Corp X156001000.0
Encompass_Preferred
Nutritionally enriched medium for the growth of recombinant strains of E. Coli and in the preparation and transformation of competent cells. Dissolve 31g in one liter distilled water. Heat solution to boiling with continuous stirring. Sterilize by autoclaving at 121?C for 15 minutes. Final ph @ 25?C: 6.8-7.2. Ingredients: Digest Peptone: 16 g/l Yeast Extract: 10 g/l Sodium Chloride: 5 g/l
Catalog No. 50-489-139
missing translation for 'QuantityLeft'
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.