Learn More
Sklar Instruments Scissors, Spencer Stitch Scissors, Sklar 3-1/2 in., Blunt/Blunt, Straight, Smooth, Delicate, Stainless Steel, EA.
Supplier: Sklar Instruments 222834
Scissors, Spencer Stitch Scissors, Sklar, 3-1/2 in., Blunt/Blunt, Straight, Smooth, Delicate, Stainless Steel, EA. Sklar Spencer Stitch Scissors are primarily used for suture removal. These scissors have a small hook shaped tip on one blade that slides under sutures to slightly lift them before cutting for removal. This hook also holds the suture so it does not slip off the blade of the scissors prior to cutting. They are most commonly available in lengths of 3-1/2 in. and 4-1/2 in. and therefore typically have smaller, more delicate tips that are used for removing smaller sutures. They should not be used for cutting tissue. This product is straight with smooth, blunt/blunt tips and a length of 3-1/2 inches. It also features a delicate pattern.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.