Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-(1H-Pyrazol-1-ylmethyl)aniline, 97%, Thermo Scientific™

Catalog No. p-4510728
Click to view available options
Quantity:
250 mg
1 g
5 g

Chemical Identifiers

CAS 892502-09-9
Molecular Formula C10H11N3
Molecular Weight (g/mol) 173.219
InChI Key JGTYTUGTENJXBY-UHFFFAOYSA-N
Synonym 3-1h-pyrazol-1-ylmethyl aniline, 3-1h-pyrazol-1-yl methyl aniline, 3-pyrazol-1-ylmethyl-phenylamine, 3-pyrazol-1-ylmethyl aniline, 3-1-pyrazolyl methyl aniline, 3-1h-pyrazol-1-ylmethyl phenyl amine, 3-pyrazolylmethyl phenylamine
PubChem CID 6485349
IUPAC Name 3-(pyrazol-1-ylmethyl)aniline
SMILES C1=CC(=CC(=C1)N)CN2C=CC=N2

Specifications

Assay Percent Range 97%
Quantity 250 mg
Formula Weight 173.22
Percent Purity 97%
Chemical Name or Material 3-(1H-Pyrazol-1-ylmethyl)aniline
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.