Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-(2,6-Dichlorophenyl)-5-methylisoxazole-4-carboxylic acid, 97%, Thermo Scientific™

Catalog No. SB00572EA
Click to view available options
Quantity:
10 g

Chemical Identifiers

CAS 3919-76-4
Molecular Formula C11H7Cl2NO3
Molecular Weight (g/mol) 272.08
MDL Number MFCD00051658
InChI Key WQXUUMUOERZZAE-UHFFFAOYSA-N
Synonym 3-2,6-dichlorophenyl-5-methylisoxazole-4-carboxylic acid, 5-methyl-3-2',6'-dichlorophenyl-4-isoxazolecarboxylic acid, 4-isoxazolecarboxylic acid, 3-2,6-dichlorophenyl-5-methyl, 3-2,6-dichlorophenyl-5-methyl-1,2-oxazole-4-carboxylic acid, 3-2,6-dichlorophenyl-5-methyl-4-isoxazolecarboxylic acid, pubchem8684, dicloxacillin impurity d, acmc-1cmb3, pyridinium hydroxy propyl sulphobetaine; pps-oh, 3-2,6-dichlorophenyl-5-methylisoxazol-4-carboxylic acid
PubChem CID 77521
IUPAC Name 3-(2,6-dichlorophenyl)-5-methyl-1,2-oxazole-4-carboxylic acid
SMILES CC1=C(C(O)=O)C(=NO1)C1=C(Cl)C=CC=C1Cl

Specifications

Assay Percent Range 97%
Packaging Amber glass bottle
Quantity 10 g
Formula Weight 272.09
Percent Purity 97%
Chemical Name or Material 3-(2, 6-Dichlorophenyl)-5-methylisoxazole-4-carboxylic acid
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.