Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3(2H)-Pyridazinone 96.0+%, TCI America™
SDP

Catalog No. p-7135289 Shop All Tokyo Chemical Industry (TCI) Products
Click to view available options
Quantity:
5 g
25 g

Chemical Identifiers

CAS 504-30-3
Molecular Formula C4H4N2O
Molecular Weight (g/mol) 96.089
MDL Number MFCD00099605
InChI Key AAILEWXSEQLMNI-UHFFFAOYSA-N
Synonym 3 2h-pyridazinone, pyridazin-3 2h-one, 2h-pyridazin-3-one, 3-hydroxypyridazine, 3-pyridazinol, pyridazin-3-ol, pyridazinone, 2,3-dihydropyridazin-3-one, 3-pyridazinone, 6-pyridazinone
PubChem CID 68153
IUPAC Name 1H-pyridazin-6-one
SMILES C1=CC(=O)NN=C1

Specifications

Melting Point 102°C
Color White-Yellow
Boiling Point 101°C
Quantity 5 g
Formula Weight 96.09
Percent Purity ≥96.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 3(2H)-Pyridazinone
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.