Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific Chemicals 3,3'-Diaminobenzidine, 99%

Catalog No. AC112080250
Click to view available options
Quantity:
Packaging:

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS

91-95-2

Molecular Formula

C12H14N4

Molecular Weight (g/mol)

214.27

MDL Number

MFCD00007725

InChI Key

HSTOKWSFWGCZMH-UHFFFAOYSA-N

Synonym

3,3'-diaminobenzidine, 3,3',4,4'-tetraaminobiphenyl, 3,3'-diaminobenzidene, 3,3',4,4'-diphenyltetramine, 3,3',4,4'-tetraminobiphenyl, 3,3',4,4'-tetraaminodiphenyl, 1,1'-biphenyl-3,3',4,4'-tetramine, biphenyl-3,3',4,4'-tetrayltetraamine, unii-2rv4t6khqi, 4-3,4-diaminophenyl benzene-1,2-diamine

PubChem CID

7071

IUPAC Name

4-(3,4-diaminophenyl)benzene-1,2-diamine

SMILES

C1=CC(=C(C=C1C2=CC(=C(C=C2)N)N)N)N

Specifications

CAS Min %

98.5

CAS Max %

100.0

Melting Point

176.0°C to 180.0°C

Flash Point

>200°C

Assay Percent Range

99%

Packaging

Glass bottle

Quantity

25 g

Beilstein

13, 340

Solubility Information

Solubility in water: 0.55g/L in water (20°C); 25g/L in water (95°C).

Formula Weight

214.27

Percent Purity

99%

Chemical Name or Material

3, 3'-Diaminobenzidine, 99%

Hazard Category

GHS Signal Word: Danger

Hazard Statement

GHS H Statement:
Suspected of causing genetic defects.
May cause cancer.

Precautionary Statement

GHS P Statement:
Use personal protective equipment as required.
Obtain special instructions before use.
IF exposed or concerned: Get medical advice/attention.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

EINECSNumber

202-110-6

RTECSNumber

DV8750000

TSCA

TSCA

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.