Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3,4-Benzopyrene (purified by sublimation) 95.0+%, TCI America™
SDP

Catalog No. p-7118343 Shop All Tokyo Chemical Industry (TCI) Products
Click to view available options
Quantity:
100 mg
1 g

Chemical Identifiers

CAS 50-32-8
Molecular Formula C20H12
Molecular Weight (g/mol) 252.32
MDL Number MFCD00003602
InChI Key FMMWHPNWAFZXNH-UHFFFAOYSA-N
Synonym benzo a pyrene, 3,4-benzopyrene, benzo pqr tetraphene, 3,4-benzpyrene, benzpyrene, benzo def chrysene, 6,7-benzopyrene, benz a pyrene, 3,4-benz a pyrene
PubChem CID 2336
ChEBI CHEBI:29865
IUPAC Name pentacyclo[10.6.2.0²,⁷.0⁹,¹⁹.0¹⁶,²⁰]icosa-1(18),2,4,6,8,10,12,14,16,19-decaene
SMILES C1=CC=C2C(=C1)C=C1C=CC3=CC=CC4=CC=C2C1=C34

Specifications

Melting Point 176°C
Boiling Point 496°C
Quantity 100 mg
UN Number 3077
Formula Weight 252.32
Percent Purity ≥95.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 3,4-Benzopyrene (purified by sublimation)
RTECSNumber DJ3675000
TSCA Yes
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.